4H-1,2,4-Triazole 95%

Supplier: Ambeed
Warning

A920810-10G A920810-1G A920810-5G
76880-880EA 109.28 USD
76880-880 76880-882 76880-884
4H-1,2,4-Triazole 95%
4H-1,2,4-Triazole

Precaución: for Research Use Only

Formula: C₂H₃N₃
MW: 69.06 g/mol
MDL Number: MFCD01941334
CAS Number: 63598-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR