2,2'-Bis(diphenylphosphino)biphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
BIPHEP, 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl
A920440-250MG
A920440-1G
A920440-100MG
A920440-5G
76880-878EA
51
USD
76880-878
77552-716
77614-642
77674-368
2,2'-Bis(diphenylphosphino)biphenyl 98%
2,2'-Bis(diphenylphosphino)biphenyl
Precaución: for Research Use Only
Formula:
C₃₆H₂₈P₂ |
MDL Number:
MFCD03094574 CAS Number: 84783-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...