4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Cyclopentylcarbamoyl)-3-fluorophenyl)boronic acid, N-Cyclopentyl 4-borono-2-fluorobenzamide, 4-(Cyclopentylcarbamoyl)-3-fluorophenylboronic acid

A914602-100MG A914602-1G A914602-250MG
76880-852EA 39 USD
76880-852 76880-854 76880-856
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid 98%
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BFNO₃
MDL Number: MFCD09475825
CAS Number: 957034-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR