Methyl 6-(aminomethyl)nicotinate hydrochloride 95%

Supplier: Ambeed
Warning

A912476-100MG A912476-250MG A912476-5G
76880-848EA 27 USD
76880-848 76880-850 77886-908
Methyl 6-(aminomethyl)nicotinate hydrochloride 95%
Methyl 6-(aminomethyl)nicotinate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MDL Number: MFCD11045450
CAS Number: 1072438-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR