4-(1-Boc-4-piperidyl)-1-butanol 98%

Supplier: Ambeed
Warning

A911366-100MG A911366-1G A911366-250MG A911366-25G A911366-5G
76880-836EA 24 USD
76880-836 76880-838 76880-840 76880-842 77614-628
4-(1-Boc-4-piperidyl)-1-butanol 98%
4-(1-Boc-4-piperidyl)-1-butanol

Precaución: for Research Use Only

Formula: C₁₄H₂₇NO₃
MDL Number: MFCD03265708
CAS Number: 142355-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR