1-(2-Pyridyl)cyclopropylamine Dihydrochloride 98%

Supplier: Ambeed
Warning

A910934-1G A910934-250MG A910934-100MG
76880-834EA 150 USD
76880-834 77674-348 77884-857
1-(2-Pyridyl)cyclopropylamine Dihydrochloride 98%
1-(2-Pyridyl)cyclopropylamine Dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂
MDL Number: MFCD09028134
CAS Number: 1215107-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR