5-Fluoro-2-(methylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoro-2-(methylcarbamoyl)phenyl)boronic acid

A909037-100MG A909037-1G A909037-250MG
76880-820EA 52 USD
76880-820 76880-822 76880-824
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid 98%
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
MDL Number: MFCD09878343
CAS Number: 957062-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR