Methyl 2-(1-methyl-1H-pyrazol-4-yl)acetate 95%

Supplier: Ambeed
Warning

A904943-100MG A904943-250MG A904943-1G A904943-5G
76880-804EA 45 USD
76880-804 76880-806 77595-456 77884-849
Methyl 2-(1-methyl-1H-pyrazol-4-yl)acetate 95%
Methyl 2-(1-methyl-1H-pyrazol-4-yl)acetate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD17267695
CAS Number: 1248548-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR