5'-Chloro-2'-hydroxy-3'-nitroacetophenone 95%

Supplier: Ambeed
Danger

Synonyms: 1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene, 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone

A903354-100G A903354-25G A903354-500G
76880-798EA 18 USD
76880-798 76880-800 76880-802
5'-Chloro-2'-hydroxy-3'-nitroacetophenone 95%
5'-Chloro-2'-hydroxy-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
MW: 215.59
MDL Number: MFCD00191921
CAS Number: 84942-40-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR