(4-Bromobutyl)boronic acid 95+%
Supplier: Ambeed
|
Warning
|
A901148-100MG
A901148-1G
A901148-250MG
A901148-5G
76880-788EA
46
USD
76880-788
76880-790
76880-792
77886-898
(4-Bromobutyl)boronic acid 95+%
(4-Bromobutyl)boronic acid
Precaución: for Research Use Only
Formula:
C₄H₁₀BBrO₂ |
MDL Number:
MFCD10000895 CAS Number: 61632-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...