(2S,4S)-1-(tert-Butoxycarbonyl)-4-methoxypyrrolidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-cis-Hyp(Me)-OH

A900231-250MG A900231-1G A900231-5G
76880-782EA 31 USD
76880-782 77674-338 77884-847
(2S,4S)-1-(tert-Butoxycarbonyl)-4-methoxypyrrolidine-2-carboxylic acid 98%
(2S,4S)-1-(tert-Butoxycarbonyl)-4-methoxypyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
MDL Number: MFCD11501217
CAS Number: 83623-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR