3-Morpholino-1-[4-(2-oxo-1-piperidyl)phenyl]-5,6-dihydropyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A893718-10G A893718-1G A893718-25G A893718-5G A893718-100G
76880-750EA 38.2 USD
76880-750 76880-752 76880-754 76880-756 77674-324
3-Morpholino-1-[4-(2-oxo-1-piperidyl)phenyl]-5,6-dihydropyridin-2(1H)-one 95%
3-Morpholino-1-[4-(2-oxo-1-piperidyl)phenyl]-5,6-dihydropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O₃
MW: 355.43 g/mol
MDL Number: MFCD19440881
CAS Number: 545445-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR