4-Nitrodiphenylamine 98%

Supplier: Ambeed
Warning

A886768-1G A886768-250MG
76880-728EA 32 USD
76880-728 76880-730
4-Nitrodiphenylamine 98%
4-Nitrodiphenylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD00007301
CAS Number: 836-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR