2-Methyl-4-pyridineboronic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2-Methylpyridin-4-yl)boronic acid hydrochloride

A881431-10G A881431-1G A881431-25G A881431-5G
76880-720EA 120.95 USD
76880-720 76880-722 76880-724 76880-726
2-Methyl-4-pyridineboronic acid hydrochloride 95%
2-Methyl-4-pyridineboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BClNO₂
MW: 173.4100 g/mol
MDL Number: MFCD08277258
CAS Number: 861905-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR