4-Bromo-5-methoxybenzofuran 95+%

Supplier: Ambeed
Warning

A880722-100MG A880722-250MG
76880-716EA 79 USD
76880-716 76880-718
4-Bromo-5-methoxybenzofuran 95+%
4-Bromo-5-methoxybenzofuran

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.05 g/mol
MDL Number: MFCD23705127
CAS Number: 36826-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR