1-(4-Iodophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A880509-250MG A880509-5G
76880-714EA 15.92 USD
76880-714 77614-582
1-(4-Iodophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one 95%
1-(4-Iodophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₇IN₂O₂
MW: 384.212 g/mol
MDL Number: MFCD18072445
CAS Number: 473927-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR