4-Carboxy-3-methylphenylboronic acid, pinacol ester 96%

Supplier: Ambeed
Warning

A879732-100MG A879732-1G A879732-250MG A879732-5G
76880-708EA 37 USD
76880-708 76880-710 76880-712 77886-881
4-Carboxy-3-methylphenylboronic acid, pinacol ester 96%
4-Carboxy-3-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MDL Number: MFCD09037836
CAS Number: 890839-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR