tert-Butyl (3-(methylamino)propyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A876205-1G A876205-250MG A876205-5G
76880-704EA 56.23 USD
76880-704 76880-706 77614-570
tert-Butyl (3-(methylamino)propyl)carbamate hydrochloride 95%
tert-Butyl (3-(methylamino)propyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₁ClN₂O₂
MW: 224.72 g/mol
MDL Number: MFCD11101326
CAS Number: 1188264-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR