4-Bromo-N-(4-methoxybenzyl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A875461-1G A875461-250MG A875461-5G
76880-684EA 93.36 USD
76880-684 76880-686 76880-688
4-Bromo-N-(4-methoxybenzyl)benzenesulfonamide 98%
4-Bromo-N-(4-methoxybenzyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrNO₃S
MW: 356.2349 g/mol
MDL Number: MFCD01359619
CAS Number: 329939-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR