4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate 98%

Supplier: Ambeed
Danger

Synonyms: p-Cumenyl(p-tolyl)iodonium Tetrakis(pentafluorophenyl)borate, (4-Isopropylphenyl)(p-tolyl)iodonium tetrakis(perfluorophenyl)borate

A874195-1G A874195-25G A874195-5G
76880-678EA 14.86 USD
76880-678 76880-680 76880-682
4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate 98%
4-Isopropyl-4'-methyldiphenyliodonium tetrakis(pentafluorophenyl)borate

Precaución: for Research Use Only

Formula: C₄₀H₁₈BF₂₀I
MW: 1016.27 g/mol
MDL Number: MFCD03411231
CAS Number: 178233-72-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR