(6-(Methylsulfonyl)pyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

A866111-100MG A866111-1G A866111-250MG A866111-5G
76880-630EA 21 USD
76880-630 76880-632 76880-634 76880-636
(6-(Methylsulfonyl)pyridin-3-yl)boronic acid 98%
(6-(Methylsulfonyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₄S
MDL Number: MFCD12964553
CAS Number: 1088496-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR