2-Chlor-6-fluor-4-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A863859-1G A863859-250MG A863859-5G
76880-612EA 201.58 USD
76880-612 76880-614 76880-616
2-Chlor-6-fluor-4-pyridineboronic acid pinacol ester 98%
2-Chlor-6-fluor-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BClFNO₂
MW: 257.5 g/mol
MDL Number: MFCD11504972
CAS Number: 1146615-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR