2-(4-Chloro-3-methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A861924-1G A861924-250MG A861924-5G
76880-600EA 82.76 USD
76880-600 76880-602 76880-604
2-(4-Chloro-3-methoxyphenyl)acetonitrile 98%
2-(4-Chloro-3-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.62 g/mol
MDL Number: MFCD09907926
CAS Number: 13726-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR