Methyl 4-bromo-2-(cyanomethyl)benzoate 95%

Supplier: Ambeed
Warning

A854913-1G A854913-250MG A854913-5G
76880-578EA 115.64 USD
76880-578 76880-580 77614-524
Methyl 4-bromo-2-(cyanomethyl)benzoate 95%
Methyl 4-bromo-2-(cyanomethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
MDL Number: MFCD11559034
CAS Number: 1083181-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR