4-Ethoxy-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A854492-100MG A854492-1G A854492-250MG
76880-572EA 50.93 USD
76880-572 76880-574 76880-576
4-Ethoxy-2-fluoroaniline 98%
4-Ethoxy-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MW: 155.17 g/mol
MDL Number: MFCD11053973
CAS Number: 470702-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR