4-Carboxy-3-chlorophenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-3-chlorophenylboronic acid, pinacol ester, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A854224-1G A854224-250MG A854224-5G
76880-566EA 120.95 USD
76880-566 76880-568 76880-570
4-Carboxy-3-chlorophenylboronic acid pinacol ester 96%
4-Carboxy-3-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BClO₄
MW: 282.5277 g/mol
MDL Number: MFCD12546621
CAS Number: 890839-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR