[2,2'-Bipyridine]-6-carbaldehyde 95%

Supplier: Ambeed
Warning

A854114-1G A854114-250MG A854114-100MG A854114-5G
76880-562EA 255 USD
76880-562 76880-564 77674-264 77674-266
[2,2'-Bipyridine]-6-carbaldehyde 95%
[2,2'-Bipyridine]-6-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O
MDL Number: MFCD11042462
CAS Number: 134296-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR