(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylicacid amide 95%

Supplier: Ambeed
Warning

A851580-1G A851580-250MG
76880-552EA 248 USD
76880-552 77884-783
(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylicacid amide 95%
(S)-1-(2-Chloro-acetyl)-pyrrolidine-2-carboxylicacid amide

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD11845729
CAS Number: 214398-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR