2-([1,1-Biphenyl]-4-yl)-5-phenyl-1,3,4-oxadiazole 98%

Supplier: Ambeed
Warning

A850324-1G A850324-250MG A850324-5G A850324-25G
76880-546EA 35 USD
76880-546 76880-548 76880-550 77884-781
2-([1,1-Biphenyl]-4-yl)-5-phenyl-1,3,4-oxadiazole 98%
2-([1,1-Biphenyl]-4-yl)-5-phenyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O
MDL Number: MFCD00003100
CAS Number: 852-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR