2-Ethylisothiazolidine 1,1-dioxide 98%

Supplier: Ambeed
Warning

A846234-1G A846234-5G
76880-536EA 74.27 USD
76880-536 76880-538
2-Ethylisothiazolidine 1,1-dioxide 98%
2-Ethylisothiazolidine 1,1-dioxide

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD10699691
CAS Number: 73343-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR