(5-Chloro-2-isopropoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A846104-1G A846104-250MG A846104-5G
76880-530EA 16 USD
76880-530 76880-532 76880-534
(5-Chloro-2-isopropoxyphenyl)boronic acid 98%
(5-Chloro-2-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
MDL Number: MFCD06411283
CAS Number: 352534-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR