4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(tert-Butylcarbamoyl)-3-fluorophenyl)boronic acid

A843696-100MG A843696-1G A843696-250MG A843696-5G
76880-522EA 33 USD
76880-522 76880-524 76880-526 76880-528
4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid 98%
4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MDL Number: MFCD08689525
CAS Number: 874289-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR