6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride 98%

Supplier: Ambeed
Warning

A841739-1G A841739-250MG A841739-5G
76880-516EA 12 USD
76880-516 76880-518 76880-520
6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride 98%
6-Bromo-7-methylimidazo[1,2-a]pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₈BrClN₂
MW: 247.5200 g/mol
MDL Number: MFCD09475875
CAS Number: 957035-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR