Pyrido[4,3-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

A840424-100MG A840424-1G A840424-250MG A840424-5G
76880-508EA 29.71 USD
76880-508 76880-510 76880-512 76880-514
Pyrido[4,3-d]pyrimidin-4(3H)-one 98%
Pyrido[4,3-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₅N₃O
MW: 147.13 g/mol
MDL Number: MFCD09881280
CAS Number: 16952-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR