3-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Ethyl(methyl)carbamoyl)phenyl)boronic acid, 3-[(N-Ethyl-N-methylamino)carbonyl]benzeneboronic acid

A839718-1G A839718-250MG A839718-5G
76880-498EA 94 USD
76880-498 76880-500 76880-502
3-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%
3-[Ethyl(methyl)carbamoyl]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₃
MDL Number: MFCD08056369
CAS Number: 871333-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR