1-Tosylazetidin-3-one 98%
Supplier: Ambeed
|
Warning
|
A837759-100MG
A837759-1G
A837759-250MG
A837759-5G
76880-490EA
22.28
USD
76880-490
76880-492
76880-494
76880-496
1-Tosylazetidin-3-one 98%
1-Tosylazetidin-3-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₃S |
MDL Number:
MFCD00129860 CAS Number: 76543-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...