5-Bromo-8-fluoroquinoline 98%

Supplier: Ambeed
Warning

A834610-100MG A834610-1G A834610-250MG
76880-472EA 20.16 USD
76880-472 76880-474 76880-476
5-Bromo-8-fluoroquinoline 98%
5-Bromo-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0500 g/mol
MDL Number: MFCD11855895
CAS Number: 1133115-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR