2-Benzoylbenzoic acid 98%

Supplier: Ambeed
Warning

A833753-1000G A833753-100G A833753-500G
76880-464EA 37 USD
76880-464 76880-466 76880-468 76880-470
2-Benzoylbenzoic acid 98%
2-Benzoylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₃
MDL Number: MFCD00002472
CAS Number: 85-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR