5-Bromo-N-cyclopropylnicotinamide 97%

Supplier: Ambeed
Warning

A832323-1G A832323-250MG A832323-5G
76880-454EA 60.48 USD
76880-454 76880-456 76880-458
5-Bromo-N-cyclopropylnicotinamide 97%
5-Bromo-N-cyclopropylnicotinamide

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MW: 241.09 g/mol
MDL Number: MFCD03017678
CAS Number: 385382-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR