4-(N,N-Dimethylsulfamoyl)-2-methylbenzeneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl 4-borono-3-methylbenzenesulphonamide, (4-(N,N-Dimethylsulphamoyl)-2-methylphenyl)boronic acid

A830690-1G A830690-250MG A830690-5G
76880-444EA 201.58 USD
76880-444 76880-446 76880-448
4-(N,N-Dimethylsulfamoyl)-2-methylbenzeneboronic acid 96%
4-(N,N-Dimethylsulfamoyl)-2-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₄S
MW: 243.088 g/mol
MDL Number: MFCD09475839
CAS Number: 957034-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR