1-Boc-7-methoxyindole-3-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A828816-100MG A828816-1G A828816-250MG A828816-5G
76880-434EA 135 USD
76880-434 76880-436 76880-438 76880-440
1-Boc-7-methoxyindole-3-boronic acid pinacol ester 95%
1-Boc-7-methoxyindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₈BNO₅
MDL Number: MFCD14155739
CAS Number: 1218790-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR