5-Fluoro-2-formylbenzoic acid 95%

Supplier: Ambeed
Warning

A827852-1G A827852-5G A827852-100MG
76880-432EA 118 USD
76880-432 77573-640 77886-818
5-Fluoro-2-formylbenzoic acid 95%
5-Fluoro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₃
MDL Number: MFCD11053106
CAS Number: 920481-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR