3-Bromo-5-(tert-butyldimethylsilyloxy)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)(tert-butyl)dimethylsilane

A827142-100MG A827142-1G A827142-250MG A827142-5G
76880-422EA 37.14 USD
76880-422 76880-424 76880-426 76880-428
3-Bromo-5-(tert-butyldimethylsilyloxy)phenylboronic acid pinacol ester 95%
3-Bromo-5-(tert-butyldimethylsilyloxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₃₀BBrO₃Si
MW: 413.2300 g/mol
MDL Number: MFCD12546591
CAS Number: 1218789-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR