(6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine fumarate 95%

Supplier: Ambeed
Warning

A825510-100G A825510-1G A825510-25G A825510-500G A825510-5G
76880-412EA 212.18 USD
76880-412 76880-414 76880-416 76880-418 76880-420
(6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine fumarate 95%
(6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine fumarate

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₂O₄
MW: 340.37 g/mol
MDL Number: MFCD11865145
CAS Number: 127785-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR