5-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-hydroxyphenylboronic acid pinacol ester, Phenol, 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A825477-1G A825477-250MG
76880-408EA 35.01 USD
76880-408 76880-410
5-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 97%
5-Fluoro-2-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
MDL Number: MFCD16994352
CAS Number: 779331-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR