3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-boronocinnamate, (E)-(3-(3-Ethoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A821976-1G A821976-250MG A821976-5G
76880-394EA 79 USD
76880-394 76880-396 76880-398
3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98% stabilised
3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₄
MDL Number: MFCD08689480
CAS Number: 913835-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR