2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine

A818867-1G A818867-250MG A818867-5G
76880-378EA 63 USD
76880-378 76880-380 76880-382
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester 98%
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₃NO₂
MW: 307.5000 g/mol
MDL Number: MFCD13195760
CAS Number: 1218790-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR