3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2(1H)-one

A818073-100MG A818073-1G A818073-250MG
76880-360EA 17 USD
76880-360 76880-362 76880-364
3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one 97%
3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD15143623
CAS Number: 1215205-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR