(2-(N-Benzylsulfamoyl)-5-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A815787-1G A815787-250MG A815787-5G
76880-346EA 152.01 USD
76880-346 76880-348 76880-350
(2-(N-Benzylsulfamoyl)-5-methylphenyl)boronic acid 97%
(2-(N-Benzylsulfamoyl)-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₆BNO₄S
MDL Number: MFCD11053856
CAS Number: 1072946-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR