[2-(Aminomethyl)phenyl]methanamine dihydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: 1,2-Phenylenedimethanamine dihydrochloride

A814121-250MG A814121-1G
76880-344EA 20 USD
76880-344 77614-456
[2-(Aminomethyl)phenyl]methanamine dihydrochloride 96%
[2-(Aminomethyl)phenyl]methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂N₂·2ClH
MDL Number: MFCD00182462
CAS Number: 21294-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR